Skip to Content
MilliporeSigma
SNC80

SNC80

SNC80
Synonym(s):
(+)-4-[(αR)-α-((2S,5R)-4-Allyl-2,5-dimethyl-1-piperazinyl)-3-methoxybenzyl]-N,N-diethylbenzamide, SNC 80, SNC-80, SNC80
Empirical Formula (Hill Notation):
C28H39N3O2
CAS Number:
Molecular Weight:
449.63

Products

Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing