(S)-Methyl-2-(2-chlorophenyl)-2-((2-(thiophen-2-yl)ethyl)amino)acetate hydrochloride
Empirical Formula (Hill Notation):
C15H16ClNO2S·HCl
CAS Number:
Molecular Weight:
346.27
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1