PPh3 Pd G2
Synonym(s):
Chloro(triphenylphosphine) [2-(2′-amino-1,1′-biphenyl)]palladium(II), Triphenylphosphine Pd G2
Empirical Formula (Hill Notation):
C30H25ClNPPd
CAS Number:
Molecular Weight:
572.37
Products
Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing