Skip to Content
MilliporeSigma
Potassium cyanide-13C,15N 99 atom % 13C, 98 atom % 15N

Potassium cyanide-13C,15N

Potassium cyanide-13C,15N 99 atom % 13C, 98 atom % 15N
Synonym(s):
13C,15N Labeled KCN, 13C,15N Labeled potassium cyanide
Linear Formula:
K13C15N
CAS Number:
Molecular Weight:
67.10

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing