Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Platinum-rhodium alloy, Pt94Rh6
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF10165216
Platinum-rhodium alloy, Pt94Rh6,
2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF65982954
Platinum-rhodium alloy, Pt94Rh6,
1m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF36081323
Platinum-rhodium alloy, Pt94Rh6,
1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF05088652
Platinum-rhodium alloy, Pt94Rh6,
0.5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF22065780
Platinum-rhodium alloy, Pt94Rh6,
5m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF88532286
Platinum-rhodium alloy, Pt94Rh6,
0.5m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF99767211
Platinum-rhodium alloy, Pt94Rh6,
0.2m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF07857681
Platinum-rhodium alloy, Pt94Rh6,
2m wire, 0.1mm diameter, annealed
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More