Phosphomolybdic acid
Synonym(s):
Phosphomolybdic acid hydrate, Molybdophosphoric acid
Linear Formula:
H3[P(Mo3O10)4] · xH2O
CAS Number:
Molecular Weight:
1825.25 (anhydrous basis)
Products
Brand
Formula Weight
Showing 1-1 of 1
Compare
Product Number
Description
Pricing