
Phenyl-α-MIDA-boryl aldehyde

Synonym(s):
(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)(phenyl)acetaldehyde
Empirical Formula (Hill Notation):
C13H14BNO5
CAS Number:
Molecular Weight:
275.06
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1