Skip to Content
MilliporeSigma
PD 169316 ≥98% (HPLC), solid

PD 169316

PD 169316 ≥98% (HPLC), solid
Synonym(s):
4-(4-Fluorophenyl)-2-(4-nitrophenyl)-5-(4-pyridyl)-1H-imidazole, PD 169316
Empirical Formula (Hill Notation):
C20H13FN4O2
CAS Number:
Molecular Weight:
360.34

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment