Skip to Content
MilliporeSigma
PAC-1 ≥98% (HPLC)

PAC-1

PAC-1 ≥98% (HPLC)
Synonym(s):
(4-Benzylpiperazino)acetic acid (3-allyl-2-hyroxybenzylidene)hydrazide, 1-Piperazineacetic acid, 2-(phenylmethyl)-,[[2-hydroxy-3-(2-propenyl)phenyl]methylene]hyrrazide
Empirical Formula (Hill Notation):
C23H28N4O2
CAS Number:
Molecular Weight:
392.49

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing