
Omeprazole sulfide 5-carboxylic acid

Synonym(s):
4-Methoxy-6-{[(5-methoxy-1H-benzimidazol-2-yl)thio]methyl}-5-methyl-3-pyridinecarboxylic acid
Empirical Formula (Hill Notation):
C17H17N3O4S
CAS Number:
Molecular Weight:
359.40
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing