Skip to Content
MilliporeSigma
O-(4-Nitrobenzoyl)hydroxylamine 98%, ≥97.5% (HPLC)

O-(4-Nitrobenzoyl)hydroxylamine

O-(4-Nitrobenzoyl)hydroxylamine 98%, ≥97.5% (HPLC)
Synonym(s):
NbzONH2
Empirical Formula (Hill Notation):
C7H6N2O4
CAS Number:
Molecular Weight:
182.13

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing