
N-Desmethylclomipramine-D3 hydrochloride solution

Empirical Formula (Hill Notation):
C18H18D3ClN2 · HCl
CAS Number:
Molecular Weight:
340.31
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing