Skip to Content
MilliporeSigma
NaCl Solution bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Blue screw cap with 3 loci, pack of 10 bottles

NaCl Solution

NaCl Solution bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Blue screw cap with 3 loci, pack of 10 bottles

Products

Product Category
Brand
Physical Form
Agency
Chemical Composition
Capacity (mL)
Volume (ml)
Quality Segment
Closure Type
Application
Bottle material
Vial Bottle Material
Package Size
Sterility
Sterilization Method
Technique