Skip to Content
MilliporeSigma
N-(5-Aminopentyl)biotinamide trifluoroacetate salt ≥98% (TLC), solid

N-(5-Aminopentyl)biotinamide trifluoroacetate salt

N-(5-Aminopentyl)biotinamide trifluoroacetate salt ≥98% (TLC), solid
Synonym(s):
Biotin–cadaverine
Empirical Formula (Hill Notation):
C17H29F3N4O4S
Molecular Weight:
442.50

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing