![Monoclonal Anti-CD34-APC antibody produced in mouse clone 4H11[APG], purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/305/209/2c60cb51-14e3-48f2-ad1d-c8c23004cbb0/640/2c60cb51-14e3-48f2-ad1d-c8c23004cbb0.jpg)
Monoclonal Anti-CD34-APC antibody produced in mouse
![Monoclonal Anti-CD34-APC antibody produced in mouse clone 4H11[APG], purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/305/209/2c60cb51-14e3-48f2-ad1d-c8c23004cbb0/640/2c60cb51-14e3-48f2-ad1d-c8c23004cbb0.jpg)
Synonym(s):
Anti-Mucosialin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2