
Monoclonal Anti-B3GAT3, (C-terminal) antibody produced in mouse

Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing