Skip to Content
MilliporeSigma
MMP 2 Substrate, fluorogenic

MMP-2 Substrate, Fluorogenic

MMP 2 Substrate, fluorogenic
Synonym(s):
MMP-2 Substrate, Fluorogenic
Empirical Formula (Hill Notation):
C49H68N14O15
Molecular Weight:
1093.15

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Chemical Composition
Quality Segment