
Memantine Related Compound G

Synonym(s):
3-Methyladamantan-1-amine hydrochloride
Empirical Formula (Hill Notation):
C11H19N · HCl
CAS Number:
Molecular Weight:
201.74
Products
Shipping
Formula Weight
Physical Form
Agency
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing