![Meclozine dihydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/226/269/5f06ad9d-ab2b-4ca1-bc65-5231ba06d8dc/640/5f06ad9d-ab2b-4ca1-bc65-5231ba06d8dc.png)
Meclozine dihydrochloride
![Meclozine dihydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/226/269/5f06ad9d-ab2b-4ca1-bc65-5231ba06d8dc/640/5f06ad9d-ab2b-4ca1-bc65-5231ba06d8dc.png)
Synonym(s):
Meclizine dihydrochloride, 1-([4-Chlorophenyl]phenylmethyl)-4-([3-methylphenyl]methyl)piperazine
Empirical Formula (Hill Notation):
C25H27ClN2 · 2HCl
CAS Number:
Molecular Weight:
463.87
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing