Skip to Content
MilliporeSigma
Maltyl isobutyrate ≥98%, FG

Maltyl isobutyrate

Maltyl isobutyrate ≥98%, FG
Synonym(s):
Maltol isobutyrate
Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing