
Maduramicin ammonium

Synonym(s):
Maduramycin ammonium
Empirical Formula (Hill Notation):
C47H83NO17
CAS Number:
Molecular Weight:
934.16
Products
Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1