Skip to Content
MilliporeSigma
M9, Minimal Salts, 5X powder, minimal microbial growth medium

M9, Minimal Salts, 5X

M9, Minimal Salts, 5X powder, minimal microbial growth medium

Products

Shipping
Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Sterilization Method
Showing 1-2 of 2