Skip to Content
MilliporeSigma
Isobutyl alcohol natural, ≥99%, FCC, FG

Isobutyl alcohol

Isobutyl alcohol natural, ≥99%, FCC, FG
Synonym(s):
2-Methyl-1-propanol, Isobutanol, Isobutyl alcohol
Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing