
Ibuprofen Impurity G

Synonym(s):
(1RS,4RS)-7-(2-Methylpropyl)-1-[4-(2-methylpropyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic acid
Empirical Formula (Hill Notation):
C26H32O4
CAS Number:
Molecular Weight:
408.53
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing