
Hydrocortisone Impurity K

Synonym(s):
17-Hydroxy-3,20-dioxopregn-4-en-21-yl acetate
Empirical Formula (Hill Notation):
C23H32O5
CAS Number:
Molecular Weight:
388.50
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing