Skip to Content
MilliporeSigma
Human MMP-3 ELISA Kit for serum, plasma, cell culture supernatant and urine

Human MMP-3 ELISA Kit

Human MMP-3 ELISA Kit for serum, plasma, cell culture supernatant and urine

Products

Product Category
Brand
Gene Alias
Detection Method
Package Size
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing