Skip to Content
MilliporeSigma
Guanidine-13C,15N3 hydrochloride 98 atom % 15N, 99 atom % 13C

Guanidine-13C,15N3 hydrochloride

Guanidine-13C,15N3 hydrochloride 98 atom % 15N, 99 atom % 13C
Linear Formula:
H15N13C(15NH2)2 · HCl
CAS Number:
Molecular Weight:
99.50

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing