
Glycopyrrolate Related Compound B

Synonym(s):
1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate, 1-Methylpyrrolidin-3-yl-2-cyclopentyl-2-hydroxy-2-phenylacetate, 1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate hydrochloride
Empirical Formula (Hill Notation):
C18H25NO3
CAS Number:
Molecular Weight:
303.40
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2