Skip to Content
MilliporeSigma
Glycine-13C2,15N ethyl ester hydrochloride 98 atom % 15N, 99 atom % 13C

Glycine-13C2,15N ethyl ester hydrochloride

Glycine-13C2,15N ethyl ester hydrochloride 98 atom % 15N, 99 atom % 13C
Synonym(s):
Ethyl glycinate-13C2,15N hydrochloride
Linear Formula:
H215N13CH213COOCH2CH3·HCl
Molecular Weight:
142.56

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing