
Ginsenoside Rb3

Synonym(s):
(3β,12β)-3-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-20-yl 6-O-β-D-xylopyranosyl-β-D-glucopyranoside, Gypenoside IV
Empirical Formula (Hill Notation):
C53H90O22
CAS Number:
Molecular Weight:
1079.27
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1