Skip to Content
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Frozen Human Systemic Lupus Erythematosus (SLE) PBMC
Products
Brand
Sigma-Aldrich (4)
Biological Source
Manufacturer
Physical Form
Tissue
Quality Segment
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
HUMANPBMC-0002181
Frozen Human Systemic Lupus Erythematosus (SLE) PBMC,
human peripheral blood (mononuclear cells)
Expand
View Pricing
HUMANPBMC-0002179
Frozen Human Systemic Lupus Erythematosus (SLE) PBMC,
human peripheral blood (mononuclear cells)
Expand
View Pricing
HUMANPBMC-0002180
Frozen Human Systemic Lupus Erythematosus (SLE) PBMC,
human peripheral blood (mononuclear cells)
Expand
View Pricing
HUMANPBMC-0002612
Frozen Human Systemic Lupus Erythematosus (SLE) PBMC,
human peripheral blood (mononuclear cells)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All