
Fmoc-Met-OPfp

Synonym(s):
Fmoc-Met-OPfp
Empirical Formula (Hill Notation):
C26H20F5NO4S
CAS Number:
Molecular Weight:
537.50
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1