
Fmoc-Ala-OH-2-13C

Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-alanine-2-13C, L-Alanine-2-13C, N-Fmoc
Linear Formula:
CH313CH(NH-Fmoc)CO2H
Molecular Weight:
312.32
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Functional Group
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1