
Fluorescein sodium (C.I. 45350)

Synonym(s):
Fluorescein sodium (C.I. 45350)
Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Agency
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing