
Ethylenediaminetetraacetic-d12 acid

Synonym(s):
EDTA-d12
Linear Formula:
(HO2CCD2)2N(CD2)2N(CD2CO2H)2
CAS Number:
Molecular Weight:
304.32
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing