Skip to Content
MilliporeSigma
Ethyl cinnamate natural, ≥95%, FG

Ethyl cinnamate

Ethyl cinnamate natural, ≥95%, FG
Synonym(s):
3-Phenyl-2-propenoic acid ethyl ester, Ethyl 3-phenyl-2-propenoate, NSC 6773
Linear Formula:
C6H5CH=CHCOOC2H5
CAS Number:
Molecular Weight:
176.21
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing