Skip to Content
MilliporeSigma
Ethyl 3-methyl-2-oxopentanoate ≥95%

Ethyl 3-methyl-2-oxopentanoate

Ethyl 3-methyl-2-oxopentanoate ≥95%
Empirical Formula (Hill Notation):
C8H14O3
CAS Number:
Molecular Weight:
158.19

Products

Brand
Formula Weight
Purity
Agency
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing