Skip to Content
MilliporeSigma
Ethyl 3-(furan-2-yl)propionate ≥98%, FG

Ethyl 3-(furan-2-yl)propionate

Ethyl 3-(furan-2-yl)propionate ≥98%, FG
Synonym(s):
Ethyl 3-(2-furyl)propanoate
Empirical Formula (Hill Notation):
C9H12O3
CAS Number:
Molecular Weight:
168.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing