
Ethyl 2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate

Empirical Formula (Hill Notation):
C13H12ClNO2S
CAS Number:
Molecular Weight:
281.76
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing