Skip to Content
MilliporeSigma
DAF-FM ≥98% (HPLC)

DAF-FM

DAF-FM ≥98% (HPLC)
Synonym(s):
4-Amino-5-methylamino-2′,7′-difluorescein, Diaminofluorescein-FM
Empirical Formula (Hill Notation):
C21H14F2N2O5
CAS Number:
Molecular Weight:
412.34

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing