![Cyclopentolate for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/148/557/f5a2d12e-efb9-48cb-8ebf-59aa40a388de/640/f5a2d12e-efb9-48cb-8ebf-59aa40a388de.png)
Cyclopentolate for system suitability
![Cyclopentolate for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/148/557/f5a2d12e-efb9-48cb-8ebf-59aa40a388de/640/f5a2d12e-efb9-48cb-8ebf-59aa40a388de.png)
Synonym(s):
Cyclopentolate hydrochloride, 2-Dimethylaminoethyl 1-hydroxy-α-phenylcyclopentaneacetate
Empirical Formula (Hill Notation):
C17H25NO3 · HCl
CAS Number:
Molecular Weight:
327.85
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing