
Conjugated (9Z,11E)-Linoleic acid

Synonym(s):
(9Z,11E)-9,11-Octadecadienoic acid, 9Z,11E-CLA, Bovinic acid, Linoleic acid (9-cis, 11-trans)
Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
Products
Product Category
Brand
Biological Source
Formula Weight
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1