
Cimetidine Impurity B

Synonym(s):
Methyl 3-cyano-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl]ethyl]carbamimidate
Empirical Formula (Hill Notation):
C10H15N5SO · HCl
CAS Number:
Molecular Weight:
289.78
Products
Shipping
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing