Skip to Content
MilliporeSigma
CI 976 >98% (HPLC), solid

CI 976

CI 976 >98% (HPLC), solid
Synonym(s):
2,2-Dimethyl-N-(2,4,6-trimethoxyphenyl)dodecanamide
Empirical Formula (Hill Notation):
C23H39NO4
CAS Number:
Molecular Weight:
393.56

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing