
Boc-Ala-OH-3,3,3-d3

Synonym(s):
N-(tert-Butoxycarbonyl)-L-alanine-3,3,3-d3, L-Alanine-3,3,3-d3, N-t-Boc derivative
Linear Formula:
CD3CH(NH-Boc)CO2H
CAS Number:
Molecular Weight:
192.23
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing