Skip to Content
MilliporeSigma
Bis(4-tert-butylphenyl)iodonium triflate electronic grade, ≥99% trace metals basis

Bis(4-tert-butylphenyl)iodonium triflate

Bis(4-tert-butylphenyl)iodonium triflate electronic grade, ≥99% trace metals basis
Synonym(s):
Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate, DtBPIT
Linear Formula:
[(CH3)3CC6H4]2ICF3SO3
CAS Number:
Molecular Weight:
542.39

Products

Shipping
Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing