Skip to Content
MilliporeSigma
BDE No 77 solution 50 μg/mL in isooctane, analytical standard

BDE No 77 solution

BDE No 77 solution 50 μg/mL in isooctane, analytical standard
Synonym(s):
3,3′,4,4′-Tetrabromodiphenyl ether solution, 3,3′,4,4′-TetraBDE, PBDE 77
Empirical Formula (Hill Notation):
C12H6Br4O
CAS Number:
Molecular Weight:
485.79
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing