Skip to Content
MilliporeSigma
Baclofen solution 500 μg/mL (90:10 MeOH:DMSO), certified reference material, ampule of 1 mL, Cerilliant®

Baclofen solution

Baclofen solution 500 μg/mL (90:10 MeOH:DMSO), certified reference material, ampule of 1 mL, Cerilliant®
Empirical Formula (Hill Notation):
C10H12ClNO2
Molecular Weight:
213.66

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique