
Anti-SIRPA antibody produced in rabbit

Synonym(s):
Anti-SHPS-1, Anti-SIRPa, Anti-BIT, Anti-CD172A, Anti-MFR, Anti-MYD-1, Anti-Signal-regulatory protein α
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing