Anti-MAFB antibody produced in rabbit
Synonym(s):
Anti-Transcription factor MafB, Anti-V-maf musculoaponeurotic fibrosarcoma oncogene homolog B, Anti-KRML, Anti-MGC43127, Anti-V-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian)
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique